Learn More
Grainger Smith Bearing Cam Follower, Heavy Stud Type, High-Alloy Steel, 1/2 in. Roller Diameter, 1 ea
Catalog No. 502770514
Encompass_Preferred
These HR Series heavy stud type cam followers feature a larger diameter stud designed for applications involving high running loads and heavy shock loads where minimizing deflection is critical. Constructed from high-alloy bearing quality steel, these needle roller bearings provide robust performance in demanding industrial environments.
  • Manufactured from high-alloy bearing quality steel
  • Designed for high running loads and heavy shock loads
  • Features a larger diameter stud for reduced deflection
  • Available in sealed or unsealed configurations
  • Operating temperature range of -30°F to 250°F
Catalog No. 50-277-0514 Supplier Grainger Supplier No. 13W767
These HR Series heavy stud type cam followers feature a larger diameter stud designed for applications involving high running loads and heavy shock loads where minimizing deflection is critical. Constructed from high-alloy bearing quality steel, these needle roller bearings provide robust performance in demanding industrial environments.
  • Manufactured from high-alloy bearing quality steel
  • Designed for high running loads and heavy shock loads
  • Features a larger diameter stud for reduced deflection
  • Available in sealed or unsealed configurations
  • Operating temperature range of -30°F to 250°F