Learn More
Grainger Smith Bearing Cam Follower, 1.375 in. Diameter, Eccentric, Sealed, 1 ea.
Catalog No. 502773317
Encompass_Preferred
This standard stud-type cam follower is designed for demanding industrial applications requiring reliable motion control. Featuring a flat roller profile and an eccentric stud configuration, this needle roller bearing facilitates precise radial alignment and load distribution. Constructed from high-alloy bearing steel with a black oxide finish, the unit is engineered for durability and consistent performance in diverse mechanical environments.
  • Equipped with a hex socket for efficient installation and adjustment
  • Features a double-sealed design to protect against contaminants
  • Offers a radial dynamic load capacity of 4,470 lbf
  • Includes a 1/2-20 UNF threaded stud for secure mounting
  • Pre-packed with general purpose EP grease for extended service life
Catalog No. 50-277-3317 Supplier Grainger Supplier No. 13W754
This standard stud-type cam follower is designed for demanding industrial applications requiring reliable motion control. Featuring a flat roller profile and an eccentric stud configuration, this needle roller bearing facilitates precise radial alignment and load distribution. Constructed from high-alloy bearing steel with a black oxide finish, the unit is engineered for durability and consistent performance in diverse mechanical environments.
  • Equipped with a hex socket for efficient installation and adjustment
  • Features a double-sealed design to protect against contaminants
  • Offers a radial dynamic load capacity of 4,470 lbf
  • Includes a 1/2-20 UNF threaded stud for secure mounting
  • Pre-packed with general purpose EP grease for extended service life