Learn More
Grainger Smith Bearing Cam Follower, Alloy Steel, Unsealed, 1-3/8 in. Roller
Catalog No. 502770592
Encompass_Preferred
These stud-type needle roller cam followers feature a threaded stud extending from the inner race to secure the bearing to a component, serving to convert rotating motion into linear motion in general-purpose applications. The hex socket design facilitates efficient installation, and the unit is constructed from alloy steel for durability in moderate load and shock scenarios.
  • Construction features AISI 52100 alloy steel material
  • Operating temperature range of -30°F to 250°F
  • Dynamic radial load capacity of 4,470 lbf and static radial load capacity of 4,300 lbf
  • Includes an oiler hole for lubrication maintenance
  • Concentric stud type with flat roller profile
  • Designed for unsealed operation
  • Compliant with ABMA standards
Catalog No. 50-277-0592 Supplier Grainger Supplier No. 13W712
These stud-type needle roller cam followers feature a threaded stud extending from the inner race to secure the bearing to a component, serving to convert rotating motion into linear motion in general-purpose applications. The hex socket design facilitates efficient installation, and the unit is constructed from alloy steel for durability in moderate load and shock scenarios.
  • Construction features AISI 52100 alloy steel material
  • Operating temperature range of -30°F to 250°F
  • Dynamic radial load capacity of 4,470 lbf and static radial load capacity of 4,300 lbf
  • Includes an oiler hole for lubrication maintenance
  • Concentric stud type with flat roller profile
  • Designed for unsealed operation
  • Compliant with ABMA standards