Learn More
Grainger Smith Bearing Cam Follower, 1 5/8 in. Diameter, Eccentric, Double Sealed, 1 ea.
Catalog No. 502773308
Encompass_Preferred
This standard stud-type cam follower is designed for industrial applications requiring precise alignment and load support. Featuring a needle roller bearing configuration and an eccentric stud, this component provides adjustable radial positioning to ensure optimal contact with the track surface. The assembly is constructed from alloy steel and includes molded acetal seals to protect against contaminants, while the internal cavity is pre-packed with high-performance general purpose EP grease for extended service life.
  • Eccentric stud design for precise radial adjustment
  • Double-sealed construction for enhanced contaminant protection
  • Alloy steel bearing and roller material for durability
  • Integrated hex socket drive for simplified installation
  • Features relube provisions accepting 3/16 in. drive type fittings
  • Suitable for operating temperatures from -30°F to 250°F
Catalog No. 50-277-3308 Supplier Grainger Supplier No. 13W756
This standard stud-type cam follower is designed for industrial applications requiring precise alignment and load support. Featuring a needle roller bearing configuration and an eccentric stud, this component provides adjustable radial positioning to ensure optimal contact with the track surface. The assembly is constructed from alloy steel and includes molded acetal seals to protect against contaminants, while the internal cavity is pre-packed with high-performance general purpose EP grease for extended service life.
  • Eccentric stud design for precise radial adjustment
  • Double-sealed construction for enhanced contaminant protection
  • Alloy steel bearing and roller material for durability
  • Integrated hex socket drive for simplified installation
  • Features relube provisions accepting 3/16 in. drive type fittings
  • Suitable for operating temperatures from -30°F to 250°F