Learn More
Grainger Smith Bearing Cam Follower, Slotted Head, Flat Roller, Sealed, High-Alloy Bearing Steel, 1 ea
Catalog No. 502770429
Encompass_Preferred
This CR-1-X stud type cam follower is designed for industrial applications requiring reliable movement under moderate running and shock loads. It features a flat-profile roller constructed from high-alloy bearing steel (AISI 52100) and utilizes a needle roller element for efficient performance. The unit includes a slotted head stud for straightforward installation and maintenance.
  • Features a 1.0000 in. roller diameter and 0.6250 in. roller width
  • Equipped with a 0.4375 in. diameter stud and 1.0000 in. stud length
  • Slotted head design for convenient drive access
  • Constructed from durable high-alloy bearing steel for high-performance applications
  • Features sealed closure to protect internal components
  • Rated with a static load capacity of 3,100 lbs. and a radial dynamic load capacity of 3,030 lbf.
Catalog No. 50-277-0429 Supplier Grainger Supplier No. 13W666
This CR-1-X stud type cam follower is designed for industrial applications requiring reliable movement under moderate running and shock loads. It features a flat-profile roller constructed from high-alloy bearing steel (AISI 52100) and utilizes a needle roller element for efficient performance. The unit includes a slotted head stud for straightforward installation and maintenance.
  • Features a 1.0000 in. roller diameter and 0.6250 in. roller width
  • Equipped with a 0.4375 in. diameter stud and 1.0000 in. stud length
  • Slotted head design for convenient drive access
  • Constructed from durable high-alloy bearing steel for high-performance applications
  • Features sealed closure to protect internal components
  • Rated with a static load capacity of 3,100 lbs. and a radial dynamic load capacity of 3,030 lbf.