Learn More
Grainger Smith Cam Follower, 2-1/2 in. Diameter, Standard Stud, 1 ea.
Catalog No. 502770279
Encompass_Preferred
This standard cam follower is a needle roller bearing designed for applications where running and shock loads are moderate. Constructed from high-quality bearing steel, this unsealed unit provides reliable performance in a variety of mechanical systems. It features a concentric stud type design with a screwdriver slot for ease of installation and maintenance.
  • Features a 2.5 in. roller diameter
  • Equipped with a 1-14 UNF thread size
  • Operates within a temperature range of -30°F to 250°F
  • Includes relube provisions that accept 3/16 in. drive type fittings, with plugs supplied for unused holes
  • Packed with general purpose EP grease for efficient lubrication
  • Static radial load capacity of 16,650 lbf
Catalog No. 50-277-0279 Supplier Grainger Supplier No. 13W676
This standard cam follower is a needle roller bearing designed for applications where running and shock loads are moderate. Constructed from high-quality bearing steel, this unsealed unit provides reliable performance in a variety of mechanical systems. It features a concentric stud type design with a screwdriver slot for ease of installation and maintenance.
  • Features a 2.5 in. roller diameter
  • Equipped with a 1-14 UNF thread size
  • Operates within a temperature range of -30°F to 250°F
  • Includes relube provisions that accept 3/16 in. drive type fittings, with plugs supplied for unused holes
  • Packed with general purpose EP grease for efficient lubrication
  • Static radial load capacity of 16,650 lbf