Learn More
Grainger Smith® Cam Follower, Needle Roller, Double Sealed, 2-1/4 in. Dia., 1 ea.
Catalog No. 502770805
Encompass_Preferred
This standard cam follower is a stud-type needle roller bearing designed for applications where running and shock loads are moderate. The unit features a flat-head design with a screwdriver slot for ease of installation and includes internal lubrication provisions with double-contact seals for extended service life.
  • Features a flat head with a screwdriver slot and a 7/8 in. concentric stud
  • Single-row needle roller bearing construction with molded acetal double-contact seals
  • Constructed from AISI 52100 alloy steel for standard-duty applications
  • Pre-packed with general-purpose EP grease and includes 3/16 in. drive-type fittings for relubrication
  • Operating temperature range from -30°F to 250°F
  • Dynamic radial load capacity of 10,370 lbf and static radial load capacity of 10,700 lbf
Catalog No. 50-277-0805 Supplier Grainger Supplier No. 13W696
This standard cam follower is a stud-type needle roller bearing designed for applications where running and shock loads are moderate. The unit features a flat-head design with a screwdriver slot for ease of installation and includes internal lubrication provisions with double-contact seals for extended service life.
  • Features a flat head with a screwdriver slot and a 7/8 in. concentric stud
  • Single-row needle roller bearing construction with molded acetal double-contact seals
  • Constructed from AISI 52100 alloy steel for standard-duty applications
  • Pre-packed with general-purpose EP grease and includes 3/16 in. drive-type fittings for relubrication
  • Operating temperature range from -30°F to 250°F
  • Dynamic radial load capacity of 10,370 lbf and static radial load capacity of 10,700 lbf