Learn More
Grainger Smith Bearing Standard Cam Follower, Eccentric Stud, 2-1/4 in. Dia., 1 ea.
Catalog No. 502773480
Encompass_Preferred
This standard cam follower is an eccentric, stud-type needle roller bearing engineered for high-performance industrial applications. It features a flat roller head design with double-sealed contact closures and a hex socket drive for secure installation. The bearing is constructed from durable alloy steel (AISI 52100) and includes a press-fit eccentric sleeve to allow for precise alignment adjustments during setup.
  • Dynamic radial load capacity of 10,370 lbf
  • Operational temperature range of -30°F to 250°F
  • Equipped with a press-fit eccentric sleeve for adjustment
  • Features 7/16 in hex socket drive and 7/8 in-14 threaded stud
  • Sealed with molded acetyl contact seals for environmental protection
  • Packed with general purpose EP grease with relube provisions
Catalog No. 50-277-3480 Supplier Grainger Supplier No. 13W760
This standard cam follower is an eccentric, stud-type needle roller bearing engineered for high-performance industrial applications. It features a flat roller head design with double-sealed contact closures and a hex socket drive for secure installation. The bearing is constructed from durable alloy steel (AISI 52100) and includes a press-fit eccentric sleeve to allow for precise alignment adjustments during setup.
  • Dynamic radial load capacity of 10,370 lbf
  • Operational temperature range of -30°F to 250°F
  • Equipped with a press-fit eccentric sleeve for adjustment
  • Features 7/16 in hex socket drive and 7/8 in-14 threaded stud
  • Sealed with molded acetyl contact seals for environmental protection
  • Packed with general purpose EP grease with relube provisions