Learn More
Grainger Smith Bearing Cam Follower, 2.75 in. Dia., Eccentric, Flat, 1 ea.
Catalog No. 502769991
Encompass_Preferred
This standard stud type cam follower is a needle roller bearing designed for industrial applications. It features an eccentric design with a hex drive for precision adjustment and a flat profile. The bearing is constructed from standard bearing steel with acetal seals and is pre-packed with general-purpose EP grease.
  • Eccentric bushing for precise radial adjustment
  • Integrated hex socket for convenient installation
  • Sealed design to protect internal components
  • Equipped with relube provisions compatible with 3/16 in. drive-type fittings
  • Finished with black oxide coating for corrosion resistance
  • Dynamic load capacity of 15,280 lb.
Catalog No. 50-276-9991 Supplier Grainger Supplier No. 13W762
This standard stud type cam follower is a needle roller bearing designed for industrial applications. It features an eccentric design with a hex drive for precision adjustment and a flat profile. The bearing is constructed from standard bearing steel with acetal seals and is pre-packed with general-purpose EP grease.
  • Eccentric bushing for precise radial adjustment
  • Integrated hex socket for convenient installation
  • Sealed design to protect internal components
  • Equipped with relube provisions compatible with 3/16 in. drive-type fittings
  • Finished with black oxide coating for corrosion resistance
  • Dynamic load capacity of 15,280 lb.