Learn More
Grainger Smith Bearing Cam Follower, Alloy Steel, Slotted, 3.5 in. Roller, 1 ea
Catalog No. 502770586
Encompass_Preferred
This standard duty stud type cam follower is designed for applications where running and shock loads are moderate. It features a needle roller bearing assembly with a slotted head for screwdriver adjustment. Constructed from alloy steel, the bearing offers robust performance in a variety of industrial environments.
  • Features a screwdriver slot on the stud head for adjustment.
  • Equipped with double-sealed closure for protection.
  • Operating temperature range from -30°F to 250°F.
  • Dynamic radial load capacity of 29,660 lbf and static radial load capacity of 31,800 lbf.
  • Finished with black oxide coating.
  • Designed with an oiler hole for lubrication.
  • Compliant with ABMA standards.
Catalog No. 50-277-0586 Supplier Grainger Supplier No. 13W702
This standard duty stud type cam follower is designed for applications where running and shock loads are moderate. It features a needle roller bearing assembly with a slotted head for screwdriver adjustment. Constructed from alloy steel, the bearing offers robust performance in a variety of industrial environments.
  • Features a screwdriver slot on the stud head for adjustment.
  • Equipped with double-sealed closure for protection.
  • Operating temperature range from -30°F to 250°F.
  • Dynamic radial load capacity of 29,660 lbf and static radial load capacity of 31,800 lbf.
  • Finished with black oxide coating.
  • Designed with an oiler hole for lubrication.
  • Compliant with ABMA standards.