Learn More
Grainger Smith Bearing Needle Roller Cam Follower, 4 in., Standard, 1 Row, Double Sealed, Slotted, 1 ea
Catalog No. 502772947
Encompass_Preferred
This standard-duty needle roller cam follower is designed for applications with moderate running and shock loads. Featuring a flat roller head and a concentric stud design, this bearing provides robust performance in demanding mechanical environments. It utilizes a screwdriver slot for installation and features a double sealed construction with contact seals for enhanced protection against contaminants.
  • Roller diameter of 4 in.
  • Roller width of 2-1/4 in.
  • Stud diameter of 1-1/2 in.
  • Dynamic radial load capacity of 35,980 lbf.
  • Static radial load capacity of 45,100 lbf.
  • Temperature range of -30°F to 250°F.
  • Slotted drive type with a 1-1/2 in. thread length.
  • Constructed from alloy steel with a molded acetyl seal.
Catalog No. 50-277-2947 Supplier Grainger Supplier No. 13W703
This standard-duty needle roller cam follower is designed for applications with moderate running and shock loads. Featuring a flat roller head and a concentric stud design, this bearing provides robust performance in demanding mechanical environments. It utilizes a screwdriver slot for installation and features a double sealed construction with contact seals for enhanced protection against contaminants.
  • Roller diameter of 4 in.
  • Roller width of 2-1/4 in.
  • Stud diameter of 1-1/2 in.
  • Dynamic radial load capacity of 35,980 lbf.
  • Static radial load capacity of 45,100 lbf.
  • Temperature range of -30°F to 250°F.
  • Slotted drive type with a 1-1/2 in. thread length.
  • Constructed from alloy steel with a molded acetyl seal.