Learn More
Grainger Smith Bearing Cam Follower, Eccentric Stud, Sealed, 4 in. Roller, 1 ea
Catalog No. 502772980
Encompass_Preferred
This standard duty needle roller cam follower is designed with a sealed, eccentric stud and a hex socket drive for precise installation and adjustment. Constructed from high-alloy bearing quality steel (AISI 52100), it provides high load capacity for demanding mechanical applications.
  • Manufactured from high-alloy bearing quality steel
  • Features a press fit eccentric sleeve for alignment
  • Equipped with double contact seals made of molded acetyl
  • Designed for operation in temperatures from -30°F to 250°F
  • Features a hex socket drive for efficient mounting
  • Static radial load capacity of 45,100 lbf
  • Dynamic radial load capacity of 24,660 lbf
Catalog No. 50-277-2980 Supplier Grainger Supplier No. 13W766
This standard duty needle roller cam follower is designed with a sealed, eccentric stud and a hex socket drive for precise installation and adjustment. Constructed from high-alloy bearing quality steel (AISI 52100), it provides high load capacity for demanding mechanical applications.
  • Manufactured from high-alloy bearing quality steel
  • Features a press fit eccentric sleeve for alignment
  • Equipped with double contact seals made of molded acetyl
  • Designed for operation in temperatures from -30°F to 250°F
  • Features a hex socket drive for efficient mounting
  • Static radial load capacity of 45,100 lbf
  • Dynamic radial load capacity of 24,660 lbf