Learn More
Grainger SMITH® Cam Follower, Heavy Duty, 5/8 in. Diameter, 1-Row, 1 ea.
Catalog No. 502771856
Encompass_Preferred
This heavy-duty needle roller cam follower is designed for demanding industrial applications requiring precision and durability. Constructed from high-quality alloy steel with acetal seals, this bearing provides reliable performance in a compact form factor. It comes pre-packed with general-purpose EP grease and includes relube provisions to accommodate 1/8 in. drive-type fittings.
  • Roller diameter of 5/8 in. with flat head design
  • One row of rolling elements for efficient load distribution
  • Static radial load capacity of 2,500 lbf and dynamic radial load capacity of 1,210 lbf
  • Operational temperature range of -30°F to 250°F
  • Features a 5/16 in.-24 stud thread size
  • Open design with a screwdriver slot for installation and adjustment
Catalog No. 50-277-1856 Supplier Grainger Supplier No. 13W768
This heavy-duty needle roller cam follower is designed for demanding industrial applications requiring precision and durability. Constructed from high-quality alloy steel with acetal seals, this bearing provides reliable performance in a compact form factor. It comes pre-packed with general-purpose EP grease and includes relube provisions to accommodate 1/8 in. drive-type fittings.
  • Roller diameter of 5/8 in. with flat head design
  • One row of rolling elements for efficient load distribution
  • Static radial load capacity of 2,500 lbf and dynamic radial load capacity of 1,210 lbf
  • Operational temperature range of -30°F to 250°F
  • Features a 5/16 in.-24 stud thread size
  • Open design with a screwdriver slot for installation and adjustment