Learn More
Grainger Smith Bearing Cam Follower, Alloy Steel, Heavy Duty, 7/8 in. Diameter, 1 ea.
Catalog No. 502771973
Encompass_Preferred
These heavy-duty cam followers are engineered for high-impact industrial applications. Featuring a large diameter stud, they are designed to accommodate heavy running loads and shock, effectively reducing stud deflection and bending. The bearing head is equipped with a screwdriver slot for ease of installation and maintenance.
  • Constructed from alloy steel for durability
  • Features an open closure and single row of needle rollers
  • Designed for heavy duty service environments
  • Temperature operating range of -30°F to 250°F
  • Dynamic radial load capacity of 2,140 lbf
  • Static radial load capacity of 41,500 lbf
  • Includes lubrication ports on the roller head and stud side
Catalog No. 50-277-1973 Supplier Grainger Supplier No. 13W770
These heavy-duty cam followers are engineered for high-impact industrial applications. Featuring a large diameter stud, they are designed to accommodate heavy running loads and shock, effectively reducing stud deflection and bending. The bearing head is equipped with a screwdriver slot for ease of installation and maintenance.
  • Constructed from alloy steel for durability
  • Features an open closure and single row of needle rollers
  • Designed for heavy duty service environments
  • Temperature operating range of -30°F to 250°F
  • Dynamic radial load capacity of 2,140 lbf
  • Static radial load capacity of 41,500 lbf
  • Includes lubrication ports on the roller head and stud side