Learn More
Grainger Smith Bearing Cam Follower, Alloy Steel, Needle Roller, 9/16 in. Roller Diameter, 1 ea
Catalog No. 502773198
Encompass_Preferred
This standard-duty cam follower features a needle roller bearing design and is engineered for applications where running and shock loads are moderate. Constructed from high-alloy bearing quality steel (AISI 52100) for durability, this stud-type bearing is suitable for various mechanical systems requiring precise radial load support.
  • Features a hex socket drive for easy installation
  • Constructed from high-alloy bearing steel
  • Designed for moderate running and shock loads
  • Operates within a temperature range of -30°F to 250°F
  • Dynamic radial load capacity of 970 lbf and static radial load capacity of 720 lbf
  • Includes a concentric stud with 1/4 in. thread length
Catalog No. 50-277-3198 Supplier Grainger Supplier No. 13W705
This standard-duty cam follower features a needle roller bearing design and is engineered for applications where running and shock loads are moderate. Constructed from high-alloy bearing quality steel (AISI 52100) for durability, this stud-type bearing is suitable for various mechanical systems requiring precise radial load support.
  • Features a hex socket drive for easy installation
  • Constructed from high-alloy bearing steel
  • Designed for moderate running and shock loads
  • Operates within a temperature range of -30°F to 250°F
  • Dynamic radial load capacity of 970 lbf and static radial load capacity of 720 lbf
  • Includes a concentric stud with 1/4 in. thread length