Learn More
Eurofins Discoverx CaM Kinase I, active, 10 µg. (14-663)
Supplier:  Eurofins Discoverx 14663
Encompass
N-terminal GST-tagged recombinant, human CaMK1 amino acids 2-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9145