Learn More
Eurofins Discoverx CaM Kinase I γ, active, 10 µg. (14-967)
Supplier:  Eurofins Discoverx 14967
Encompass
N-terminal GST-tagged recombinant, human full length CaM Kinase Iγ. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9502