Learn More
Grainger CCL Keyless Padlockable Cam Lock, Zinc Die-Cast, 10/Pk.
Catalog No. 502760190
Encompass_Preferred
This keyless, padlockable cam lock provides a secure closure solution for various cabinet and enclosure applications. Constructed from durable zinc die-cast with a silver finish, it is designed for general security use in environments such as schools, lobbies, and residential buildings.
  • Constructed from zinc die-cast for durability
  • Designed for metal or wood surfaces with a 1 1/8 in. material thickness
  • Compatible with 3/4 in. diameter mounting holes
  • Includes mounting hardware for installation
  • Padlockable design allows for flexible security control
Catalog No. 50-276-0190 Supplier Grainger Supplier No. 15X358
This keyless, padlockable cam lock provides a secure closure solution for various cabinet and enclosure applications. Constructed from durable zinc die-cast with a silver finish, it is designed for general security use in environments such as schools, lobbies, and residential buildings.
  • Constructed from zinc die-cast for durability
  • Designed for metal or wood surfaces with a 1 1/8 in. material thickness
  • Compatible with 3/4 in. diameter mounting holes
  • Includes mounting hardware for installation
  • Padlockable design allows for flexible security control