Learn More
MSC Midwest Rake CAM Set, 4 - 1/4 in Depth Settings, 2-Piece, Nylon
Catalog No. 502838988
Encompass
This CAM set is designed for use with CAM Gauge Rakes to apply materials at a precise, uniform depth. Each set contains two precision-machined CAMs, constructed from engineered nylon for abrasion resistance. This particular set provides 4 - 1/4 inch depth settings, allowing for consistent application of decorative concrete toppings, self-leveling floor coating materials, and underlayments.
  • Set includes two cams
  • 4 - 1/4 inch depth settings
  • Designed for use with gauge rakes
  • Each cam is precision-machined
  • Helps achieve desired depth settings
  • Abrasion-resistant construction
  • Constructed of engineered nylon
  • Comes with all necessary hardware for attachment
Catalog No. 50-283-8988 Supplier MSC Supplier No. 52098001
This CAM set is designed for use with CAM Gauge Rakes to apply materials at a precise, uniform depth. Each set contains two precision-machined CAMs, constructed from engineered nylon for abrasion resistance. This particular set provides 4 - 1/4 inch depth settings, allowing for consistent application of decorative concrete toppings, self-leveling floor coating materials, and underlayments.
  • Set includes two cams
  • 4 - 1/4 inch depth settings
  • Designed for use with gauge rakes
  • Each cam is precision-machined
  • Helps achieve desired depth settings
  • Abrasion-resistant construction
  • Constructed of engineered nylon
  • Comes with all necessary hardware for attachment