Learn More
ATCC CAMA-1; Breast Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502382439
Encompass
CAMA-1 is an epithelial-like cell that was isolated from a pleural effusion of a 51-year-old, White, female with breast adenocarcinoma.
Catalog No. 50-238-2439 Supplier ATCC Supplier No. HTB21
CAMA-1 is an epithelial-like cell that was isolated from a pleural effusion of a 51-year-old, White, female with breast adenocarcinoma.