Learn More
Camag Scientific Uv Cabinet 4, Consisting Of Viewing Box 4 And Uv Lamp 4, 254/366 Nm, 8 W
SDP
Catalog No. NC1154813
Encompass_Preferred
The Uv Cabinet 4 Consists Of The Uv Lamp 4 And The Viewing Box 4. The Front Of The Viewing Box 4 Is Closed With A Roller Shutter. Eyes Are Protected By The Uv Filter In The Viewing Window. The Distance Between Uv Lamp And Object Is Optimally Selected For Homogeneous Illumination And Observation Of The Entire Tlc Plate Through The Viewing Window.
Catalog No. NC1154813 Supplier Camag Scientific Supplier No. 040.2000
You must be signed in and have a business account to purchase online.
The Uv Cabinet 4 Consists Of The Uv Lamp 4 And The Viewing Box 4. The Front Of The Viewing Box 4 Is Closed With A Roller Shutter. Eyes Are Protected By The Uv Filter In The Viewing Window. The Distance Between Uv Lamp And Object Is Optimally Selected For Homogeneous Illumination And Observation Of The Entire Tlc Plate Through The Viewing Window.