Learn More
Cayman Chemical Cambendazol, Purity: ≥98%, CAS: 26097-80-3, 100 mg
SDP
Numéro de catalogue. 502983410
Encompass_Preferred
An antihelmintic agent; eradicates S. ratti adult worms from intestine and S. stercoralis larva from muscle at a dose of 50 mg/kg/day; inhibits polymerization of microtubules isolated from bovine brain (IC50 = 64.2 µM)
Numéro de catalogue. 50-298-3410 Fournisseur Cayman Chemical Code fournisseur 20841100 MG
Ajouter au panier
Ajouter au panier
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
An antihelmintic agent; eradicates S. ratti adult worms from intestine and S. stercoralis larva from muscle at a dose of 50 mg/kg/day; inhibits polymerization of microtubules isolated from bovine brain (IC50 = 64.2 µM)
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.