Learn More
Cayman Chemical Cambendazol, Purity: ≥98%, CAS: 26097-80-3, 50 mg
SDP
Catalog No. 502983538
Encompass_Preferred
An antihelmintic agent; eradicates S. ratti adult worms from intestine and S. stercoralis larva from muscle at a dose of 50 mg/kg/day; inhibits polymerization of microtubules isolated from bovine brain (IC50 = 64.2 µM)
Catalog No. 50-298-3538 Supplier Cayman Chemical Supplier No. 2084150 MG
An antihelmintic agent; eradicates S. ratti adult worms from intestine and S. stercoralis larva from muscle at a dose of 50 mg/kg/day; inhibits polymerization of microtubules isolated from bovine brain (IC50 = 64.2 µM)