Learn More
Electron Microscopy Sciences Cambridge Universal SEM Stub Interchangeable Carousel
SDP
Catalog No. 501930971
Encompass_Preferred
Universal adapter to fit Cambridge SEMs which then accepts interchangeable multi carousel, single pin stubs, or re-entrant stubs. Holds 8 pin stubs.
Catalog No. 50-193-0971 Supplier Electron Microscopy Sciences Supplier No. EMS037
Universal adapter to fit Cambridge SEMs which then accepts interchangeable multi carousel, single pin stubs, or re-entrant stubs. Holds 8 pin stubs.