Learn More
Electron Microscopy Sciences Cambridge Universal SEM Stub Interchangeable Carousel Adapter
SDP
Catalog No. 501930970
Encompass_Preferred
Universal adapter to fit Cambridge SEMs which then accepts interchangeable multi carousel, single pin stubs, or re-entrant stubs. Accepts interchangeable multi carousel, single pin stubs, or re-entrant stubs.
Catalog No. 50-193-0970 Supplier Electron Microscopy Sciences Supplier No. EMS036
Universal adapter to fit Cambridge SEMs which then accepts interchangeable multi carousel, single pin stubs, or re-entrant stubs. Accepts interchangeable multi carousel, single pin stubs, or re-entrant stubs.