Learn More
Cambridge Diagnostic Products, Inc.™ Camco Quik Stain™ and Quik Stain™ II

Description
- Both are self-buffering and intensity controlled by washing with ordinary distilled water
- CAMCO Quik Stain is a Wright's stain formulation (traditional blue coloration) and CAMCO Quik Stain II is a Wright-Giemsa stain (traditional coloration with blue-red highlights)
Order Info
Packaged in 946mL bottles
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | 946 mL |
| Boiling Point | 64°C |
| Format | Liquid |
| Type | Quik Stain |
| Color | Dark Blue |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.