Learn More
Electron Microscopy Sciences Cambridge SEM Carousel
SDP
Catalog No. 501930561
Encompass_Preferred
A standard carousel holder made to fit Cambridge SEMs. Will hold 8 pin stubs. Comes complete with hexagonal wrench.
Catalog No. 50-193-0561 Supplier Electron Microscopy Sciences Supplier No. EMS030
A standard carousel holder made to fit Cambridge SEMs. Will hold 8 pin stubs. Comes complete with hexagonal wrench.