Learn More
Med Vet International Camel Hair Brush, 9 inch
Catalog No. 502831925
Encompass_Preferred
Useful for dusting balances. Camel hair portion has full body and slightly pointed tip. Brush size 3/4? diameter by 1 1/2? long. Wooden handle. Overall length: 9?. Can also function as paint brush.
Catalog No. 50-283-1925 Supplier Med Vet International Supplier No. LB30
 Restricted Item
Add to Cart
Add to Cart
Useful for dusting balances. Camel hair portion has full body and slightly pointed tip. Brush size 3/4? diameter by 1 1/2? long. Wooden handle. Overall length: 9?. Can also function as paint brush.