Learn More
Electron Microscopy Sciences Camel Hair Brush Multi Pack
SDP
Catalog No. NC1513267
Encompass_Preferred
Useful for dusting and cleaning embedding center, balances, and sensitive instruments. Camel hair is mounted on wooden handle with metal ferrule.
Size: < 1.59 mm, 1.59 mm, 3.18 mm
Catalog No. NC1513267 Supplier Electron Microscopy Sciences Supplier No. 6557504
Useful for dusting and cleaning embedding center, balances, and sensitive instruments. Camel hair is mounted on wooden handle with metal ferrule.
Size: < 1.59 mm, 1.59 mm, 3.18 mm