Learn More
Gilson Company Inc Camel Hair Brush Set
Catalog No. NC9021680
Encompass_Preferred
Camel Hair Brush Set is a fine-quality set of 4 brushes for cleaning delicate mesh sieves. Recommended for cleaning #200 and finer woven-wire sieves and electroformed sieves. The set comes with two round-tip and two flat-tip brushes with camel hair bristles set in solvent-resistant cement.
Catalog No. NC9021680 Supplier Gilson Company Inc Supplier No. WT6
Camel Hair Brush Set is a fine-quality set of 4 brushes for cleaning delicate mesh sieves. Recommended for cleaning #200 and finer woven-wire sieves and electroformed sieves. The set comes with two round-tip and two flat-tip brushes with camel hair bristles set in solvent-resistant cement.