Learn More
Electron Microscopy Sciences Camel Hair Brush, < 1.59 mm
SDP
Catalog No. NC1513263
Encompass_Preferred
Useful for dusting and cleaning embedding center, balances, and sensitive instruments. Camel hair is mounted on wooden handle with metal ferrule.
Size: < 1.59 mm
Catalog No. NC1513263 Supplier Electron Microscopy Sciences Supplier No. 6557502
Useful for dusting and cleaning embedding center, balances, and sensitive instruments. Camel hair is mounted on wooden handle with metal ferrule.
Size: < 1.59 mm