Learn More
Ted Pella Inc Camel Hair Brushes, #1, less than 1.59mm wide, 3/pk
SDP
Supplier:  Ted Pella Inc 11859
Encompass
Useful for histology section manipulation, SEM, geology, forensic. Seamless ferrules, red enameled handles. Measurements taken just above ferrule.
Catalog No. NC1588898