Learn More
Ted Pella Inc Camel Hair Brushes, Multi Pack of 3 Brushes, #1, #2, #4
SDP
Catalog No. NC1536091
Encompass
Useful for histology section manipulation, SEM, geology, forensic. Seamless ferrules, red enameled handles. Measurements taken just above ferrule.
Catalog No. NC1536091 Supplier Ted Pella Inc Supplier No. 11870
Useful for histology section manipulation, SEM, geology, forensic. Seamless ferrules, red enameled handles. Measurements taken just above ferrule.