Learn More
Ted Pella Inc Camel Hair Brushes, #2, 1.59mm wide, 3/pk
SDP
Catalog No. NC2023341
Encompass
Useful for histology section manipulation, SEM, geology, forensic. Seamless ferrules, red enameled handles. #6 brush for larger brushing work on specimens. Measurements taken just above ferrule.
Catalog No. NC2023341 Supplier Ted Pella Inc Supplier No. 11860
Useful for histology section manipulation, SEM, geology, forensic. Seamless ferrules, red enameled handles. #6 brush for larger brushing work on specimens. Measurements taken just above ferrule.