Learn More
Electron Microscopy Sciences Camel Hair Round Brush, 9" OAL
SDP
Catalog No. 50365545
Encompass_Preferred
Useful for dusting and cleaning embedding center, balances, and sensitive instruments. Camel hair is mounted on wooden handle with metal ferrule. Round, camel hair has full body and slightly pointed tip. Brush size 3/4" diameter x 1 1/2" long.
OAL: 9"
Catalog No. 50-365-545 Supplier Electron Microscopy Sciences Supplier No. 6557501
Useful for dusting and cleaning embedding center, balances, and sensitive instruments. Camel hair is mounted on wooden handle with metal ferrule. Round, camel hair has full body and slightly pointed tip. Brush size 3/4" diameter x 1 1/2" long.
OAL: 9"