Learn More
Electron Microscopy Sciences Camel Hair Brush, 1.59 mm
SDP
Catalog No. NC1513265
Encompass_Preferred
Useful for dusting and cleaning embedding center, balances, and sensitive instruments. Camel hair is mounted on wooden handle with metal ferrule.
Size: 1.59 mm
Catalog No. NC1513265 Supplier Electron Microscopy Sciences Supplier No. 6557503
Useful for dusting and cleaning embedding center, balances, and sensitive instruments. Camel hair is mounted on wooden handle with metal ferrule.
Size: 1.59 mm