Learn More
Cayman Chemical Camellianin A, Purity: ≥95%, CAS: 109232-77-1, 5 mg
SDP
Catalog No. 502915881
Encompass_Preferred
A flavonoid with diverse biological activities; inhibits ACE by 30.16% at 500 µg/ml; induces cell cycle arrest at the G1 phase and apoptosis in HepG2 and MCF-7 cells at 200 µM; reduces hepatocyte apoptosis, as well as hepatic TNF-α, IL-6, IL-1β, and MDA levels, in a mouse model of carbon tetrachloride-induced liver injury at 30 mg/kg
Catalog No. 50-291-5881 Supplier Cayman Chemical Supplier No. 375755 MG
A flavonoid with diverse biological activities; inhibits ACE by 30.16% at 500 µg/ml; induces cell cycle arrest at the G1 phase and apoptosis in HepG2 and MCF-7 cells at 200 µM; reduces hepatocyte apoptosis, as well as hepatic TNF-α, IL-6, IL-1β, and MDA levels, in a mouse model of carbon tetrachloride-induced liver injury at 30 mg/kg