Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Camelliaside A | 135095-52-2 | 5 MG
SDP

Catalog No. 5000418318
Encompass_Preferred

Camelliaside A is a phytochemical that acts as a HER2 ligand. It has been shown to lack antifungal activity against *Rhizoctonia solani* and is applicable to breast cancer-related research.

  • Phytochemical and HER2 ligand
  • Shows no inhibitory effect on mycelial growth of *Rhizoctonia solani*
  • Applicable to breast cancer-related research
  • Off-white to light yellow solid appearance
  • Purity of 99.65%
  • Store at 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).
  • Soluble in DMSO (100 mg/mL)

Catalog No. 50-004-18318 Supplier Medchemexpress LLC Supplier No. HYN25245MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Camelliaside A is a phytochemical that acts as a HER2 ligand. It has been shown to lack antifungal activity against *Rhizoctonia solani* and is applicable to breast cancer-related research.

  • Phytochemical and HER2 ligand
  • Shows no inhibitory effect on mycelial growth of *Rhizoctonia solani*
  • Applicable to breast cancer-related research
  • Off-white to light yellow solid appearance
  • Purity of 99.65%
  • Store at 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light).
  • Soluble in DMSO (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.