Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Camelliaside A | 135095-52-2 | MFCD28988581 | 99.7% | 756.66 | C33H40O20 | 10 MG
SDP

Catalog No. 5000289167
Encompass_Preferred

Camelliaside A is a flavonoid isolated from Camellia oleifera seed pomace and supplied as a high-purity research standard for in vitro and in vivo studies.

  • Flavonoid compound with CAS number 135095-52-2.
  • Molecular formula C33H40O20 and molecular weight 756.66.
  • High purity: 99.65% (manufacturer reported).
  • Soluble in DMSO (100 mg/mL); ultrasonic assistance recommended.
  • Multiple in vivo formulations documented with ≥2.5 mg/mL solubility.
  • Storage: product at 4°C protected from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Intended for research use only.

Catalog No. 50-002-89167 Supplier Medchemexpress LLC Supplier No. HYN252410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Camelliaside A is a flavonoid isolated from Camellia oleifera seed pomace and supplied as a high-purity research standard for in vitro and in vivo studies.

  • Flavonoid compound with CAS number 135095-52-2.
  • Molecular formula C33H40O20 and molecular weight 756.66.
  • High purity: 99.65% (manufacturer reported).
  • Soluble in DMSO (100 mg/mL); ultrasonic assistance recommended.
  • Multiple in vivo formulations documented with ≥2.5 mg/mL solubility.
  • Storage: product at 4°C protected from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.