Learn More
Ted Pella Inc CAMELS HAIR BRUSH NO4 3/PK
SDP
Catalog No. NC9292167
Encompass
Brush; Camel's Hair; Tip Number 4; Useful for histology section manipulation, SEM, geology, forensic; Seamless ferrules, red enameled handles;
Catalog No. NC9292167 Supplier Ted Pella Inc Supplier No. 11862
Brush; Camel's Hair; Tip Number 4; Useful for histology section manipulation, SEM, geology, forensic; Seamless ferrules, red enameled handles;