Learn More
Braintree Scientific Camera System with Mounting Brackets
SDP
Catalog No. 501954590
Encompass
Camera System with Mounting Brackets - The camera system mounts to a standard stereotactic instrument via a standard V-mount. The camera enhances placement accuracy by allowing for a close-up view working area. CMOS Sensor: 2.0M pixels Still Image Resolution: up to 1600X1200pixels, Format: JPG, BMP Video Resolution: up to 1600X1200pixels Format: AVI Frame Rate: up to 30FPS FocalDistance: 5mm infinity Magnification: 5X- 270X Illumination: 6 whiteLEDs with adjustable brightness PC Interface: USB 2.0 USB Cable Length: 200cm **Sold for research purposes only and not for human use**
Catalog No. 50-195-4590 Supplier Braintree Scientific Supplier No. CS101
Camera System with Mounting Brackets - The camera system mounts to a standard stereotactic instrument via a standard V-mount. The camera enhances placement accuracy by allowing for a close-up view working area. CMOS Sensor: 2.0M pixels Still Image Resolution: up to 1600X1200pixels, Format: JPG, BMP Video Resolution: up to 1600X1200pixels Format: AVI Frame Rate: up to 30FPS FocalDistance: 5mm infinity Magnification: 5X- 270X Illumination: 6 whiteLEDs with adjustable brightness PC Interface: USB 2.0 USB Cable Length: 200cm **Sold for research purposes only and not for human use**