Learn More
Medchemexpress LLC Camizestrant | 2222844-89-3 | 99.3% | 476.51 | 50 MG
SDP
Catalog No. 5000348071
Encompass_Preferred
Camizestrant is a potent and orally active estrogen receptor (ER) antagonist. It is used for the study of ER+ HER2-advanced breast cancer. It exhibits anti-tumour efficacy in a dose-dependent manner in human parental MCF7 mice xenograft and decreases tumor growth in mice, with almost complete tumor growth inhibition at doses >10 mg/kg.
  • Potent and orally active estrogen receptor (ER) antagonist
  • Used for the study of ER+ HER2-advanced breast cancer
  • Exhibits anti-tumour efficacy in a dose-dependent manner
  • Decreases tumor growth in mice with significant inhibition at doses >10 mg/kg
Catalog No. 50-003-48071 Supplier Medchemexpress LLC Supplier No. HY13625550MG
Camizestrant is a potent and orally active estrogen receptor (ER) antagonist. It is used for the study of ER+ HER2-advanced breast cancer. It exhibits anti-tumour efficacy in a dose-dependent manner in human parental MCF7 mice xenograft and decreases tumor growth in mice, with almost complete tumor growth inhibition at doses >10 mg/kg.
  • Potent and orally active estrogen receptor (ER) antagonist
  • Used for the study of ER+ HER2-advanced breast cancer
  • Exhibits anti-tumour efficacy in a dose-dependent manner
  • Decreases tumor growth in mice with significant inhibition at doses >10 mg/kg