Learn More
Selleck Chemical LLC Camizestrant S8958-10MM/1ML
SDP
Catalog No. 5000024166
Encompass_Preferred
Camizestrant (AZD9833) is an orally available and selective estrogen receptor (ER) antagonist with antineoplastic activity
Catalog No. 50-000-24166 Supplier Selleck Chemical LLC Supplier No. S895810MM/1ML
Camizestrant (AZD9833) is an orally available and selective estrogen receptor (ER) antagonist with antineoplastic activity