Learn More
TARGETMOL CHEMICALS INC CAMIZESTRANT TFA 5MG
SDP
Catalog No. 502581546
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Camizestrant TFA (AZD-9833 TFA) is a potent and orally active antagonist of estrogen receptor (ER).
Catalog No. 50-258-1546 Supplier TARGETMOL CHEMICALS INC Supplier No. T11237L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Camizestrant TFA (AZD-9833 TFA) is a potent and orally active antagonist of estrogen receptor (ER).