Learn More
Cayman Chemical CaMKII-IN-1, Purity: ≥95%, CAS: 1208123-85-6, 5 mg
SDP
Catalog No. 502917344
Encompass_Preferred
A CaMKII inhibitor (IC50 = 0.063 µM); is selective for CaMKII over CaMKIV, MLCK, p38α, Akt1, and PKC (IC500s = >60, 36, 11, 30, and 21 µM, respectively)
Catalog No. 50-291-7344 Supplier Cayman Chemical Supplier No. 373035 MG
A CaMKII inhibitor (IC50 = 0.063 µM); is selective for CaMKII over CaMKIV, MLCK, p38α, Akt1, and PKC (IC500s = >60, 36, 11, 30, and 21 µM, respectively)