Learn More
Aviva Systems Biology Corp CAMK1G Antibody - N-terminal region (ARP57417_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57417_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CAMK1G. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CAMK1G. Alternate names: CLICKIII, VWS1, dJ272L16.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54001