Learn More
Carnabio Usa Inc CAMK2A(CAMK2A)/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 021095
Encompass
CaMK2(CAMK2A), Full-length, Wild type, Amino Acid 1-478, NP_741960.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0353