Learn More
Aviva Systems Biology Corp CAMK2B Antibody - C-terminal region (ARP61268_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61268_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CAMK2B. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CAMK2B. Alternate names: CAM2, CAMK2, CAMKB, MGC29528ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-318