Learn More
Carnabio Usa Inc CAMK2B(CAMK2B)/10UG/FULL-LENGT
Supplier:  Carnabio Usa Inc 0211010
Encompass
CaMK2(CAMK2B), Full-length, Wild type, Amino Acid 1-503 , NP_742078.1(iso5), Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0708