Learn More
Aviva Systems Biology Corp CAMK2D Antibody - C-terminal region (ARP61459_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61459_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CAMK2D. It was validated on Western Blot. . Alternate names: CAMKD, DKFZp686G23119, DKFZp686I2288, MGC44911ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-384